4-Chloro-2-(trifluoroacetyl)aniline hydrochloride 97%

Supplier: Ambeed
Warning

A164476-100G A164476-100MG A164476-10G A164476-500G A164476-25G A164476-5G
103844-126EA 6 USD
103844-126 77236-008 77236-010 77236-012 77236-014 77320-296 77320-298
4-Chloro-2-(trifluoroacetyl)aniline hydrochloride 97%
4-Chloro-2-(trifluoroacetyl)aniline hydrochloride

Precaución: for Research Use Only

Formula: C₈H₆Cl₂F₃NO
Melting Pt: 156…158 °C
Storage Temperature: Ambient
MDL Number: MFCD09263480
CAS Number: 173676-59-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR