1-(2-Amino-5-bromophenyl)ethanone 97%

Supplier: Ambeed
Warning

A147507-100G A147507-10G A147507-25G A147507-5G
103803-790EA 6 USD
103803-790 103841-894 77027-964 77027-966 77317-268 77317-270
1-(2-Amino-5-bromophenyl)ethanone 97%
1-(2-Amino-5-bromophenyl)ethanone

Precaución: for Research Use Only

Formula: C₈H₈BrNO
Flash Pt: 136.592 °C
Storage Temperature: Ambient
MDL Number: MFCD09834638
CAS Number: 29124-56-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR