1-(2-Amino-4-chlorophenyl)ethanone 95%

Supplier: Ambeed
Warning

A174013-100MG A174013-250MG A174013-1G A174013-5G
103806-898EA 252 USD
103806-898 103845-198 76813-310 76813-312 77301-304 77301-306
1-(2-Amino-4-chlorophenyl)ethanone 95%
1-(2-Amino-4-chlorophenyl)ethanone

Precaución: for Research Use Only

Formula: C₈H₈ClNO
MW: 169.61 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD12025287
CAS Number: 39061-72-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR