1-(2-Amino-4-bromophenyl)ethanone 97%
Supplier: Ambeed
|
Warning
|
A126471-100G
A126471-10G
A126471-1G
A126471-25G
A126471-5G
103800-814EA
29
USD
103800-814
76870-518
76870-520
77313-028
77313-030
77313-032
1-(2-Amino-4-bromophenyl)ethanone 97%
1-(2-Amino-4-bromophenyl)ethanone
Precaución: for Research Use Only
Formula:
C₈H₈BrNO MW: 214.05 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD16659002 CAS Number: 123858-51-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...