2',6'-Dimethylacetophenone 97%

Supplier: Ambeed
Warning

A127576-10G A127576-5G A127576-1G
103800-986EA 15 USD
103800-986 76688-510 76688-512 77298-230 77313-232
2',6'-Dimethylacetophenone 97%
2',6'-Dimethylacetophenone

Precaución: for Research Use Only

Formula: C₁₀H₁₂O
Boiling Pt: 225.1 °C (760 mm Hg)
Flash Pt: 85.6 °C
Storage Temperature: Ambient
MDL Number: MFCD01861394
CAS Number: 2142-76-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR