(2,4-Dimethoxybenzyl)methylamine 98%

Supplier: Ambeed
Warning

Synonyms: 1-(2,4-Dimethoxyphenyl)-N-methylmethanamine

A160454-10G A160454-250MG A160454-1G A160454-25G A160454-5G
103805-380EA 17 USD
103805-380 103843-612 76720-118 76720-120 77319-548 77319-550 77319-552
(2,4-Dimethoxybenzyl)methylamine 98%
(2,4-Dimethoxybenzyl)methylamine

Precaución: for Research Use Only

Formula: C₁₀H₁₅NO₂
MW: 181.232 g/mol
Boiling Pt: 262.1 °C (760 mm Hg)
Flash Pt: 104.6 °C
Storage Temperature: Refrigerator
MDL Number: MFCD04633426
CAS Number: 102503-23-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR