1-(2,4-Dihydroxyphenyl)butan-1-one 95%

Supplier: Ambeed
Warning

A123150-1G A123150-5G
103800-352EA 22 USD
103800-352 77297-932 77297-934
1-(2,4-Dihydroxyphenyl)butan-1-one 95%
1-(2,4-Dihydroxyphenyl)butan-1-one

Precaución: for Research Use Only

Formula: C₁₀H₁₂O₃
MW: 180.2 g/mol
Boiling Pt: 338.5 °C (760 mm Hg)
Melting Pt: 73 °C
Flash Pt: 172.7 °C
Storage Temperature: Ambient
MDL Number: MFCD01098941
CAS Number: 4390-92-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR