2',4',5'-Trichloroacetophenone 98%

Supplier: Ambeed
Warning

Synonyms: 1-(2,4,5-Trichlorophenyl)ethanone

A384244-1G A384244-250MG
103826-758EA 95 USD
103826-758 76841-142 77168-096
2',4',5'-Trichloroacetophenone 98%
2',4',5'-Trichloroacetophenone

Precaución: for Research Use Only

Formula: C₈H₅Cl₃O
MW: 223.484 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09261422
CAS Number: 13061-28-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR