2',3'-Dimethylacetophenone 95%

Supplier: Ambeed
Warning

Synonyms: 1-(2,3-Dimethylphenyl)ethanone

A138909-10G A138909-25G
103802-676EA 6 USD
103802-676 76688-944 77315-738
2',3'-Dimethylacetophenone 95%
2',3'-Dimethylacetophenone

Precaución: for Research Use Only

Formula: C₁₀H₁₂O
MW: 148.202 g/mol
Boiling Pt: 236.6 °C (760 mm Hg)
Flash Pt: 92.4 °C
Storage Temperature: Ambient
MDL Number: MFCD03844735
CAS Number: 2142-71-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR