1-(2,3-Dihydrobenzo[b][1,4]dioxin-6-yl)ethanone 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
3',4'-Ethylenedioxyacetophenone, 1,4-Benzodioxan-6-yl methyl ketone, 6-Acetyl-1,4-benzodioxane
A235779-1G
A235779-5G
A235779-10G
103811-030EA
6
USD
103811-030
103850-798
77089-816
77089-818
77615-936
1-(2,3-Dihydrobenzo[b][1,4]dioxin-6-yl)ethanone 98%
1-(2,3-Dihydrobenzo[b][1,4]dioxin-6-yl)ethanone
Precaución: for Research Use Only
Formula:
C₁₀H₁₀O₃ MW: 178.19 g/mol Boiling Pt: 313.7 °C (760 mm Hg) Melting Pt: 80…82 °C Storage Temperature: Ambient |
MDL Number:
MFCD00006823 CAS Number: 2879-20-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...