1-(2,2-Difluorobenzo[1,3]dioxol-5-yl)-cyclopropanecarboxylic acid 97%

Supplier: Ambeed
Warning

A201464-100G A201464-25G A201464-10G
103817-298EA 8 USD
103817-298 103847-938 76785-356 76785-358 77667-674
1-(2,2-Difluorobenzo[1,3]dioxol-5-yl)-cyclopropanecarboxylic acid 97%
1-(2,2-Difluorobenzo[1,3]dioxol-5-yl)-cyclopropanecarboxylic acid

Precaución: for Research Use Only

Formula: C₁₁H₈F₂O₄
MW: 242.18 g/mol
Storage Temperature: Ambient
MDL Number: MFCD13190001
CAS Number: 862574-88-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR