1-(Trifluoroacetyl)piperidine-4-carboxylic acid 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
1-(2,2,2-Trifluoroacetyl)-4-piperidinecarboxylic acid, 1-(2,2,2-Trifluoroacetyl)piperidine-4-carboxylic acid
A209085-250MG
A209085-1G
A209085-5G
103817-914EA
40
USD
103817-914
103848-600
77192-574
77303-082
77303-084
1-(Trifluoroacetyl)piperidine-4-carboxylic acid 97%
1-(Trifluoroacetyl)piperidine-4-carboxylic acid
Precaución: for Research Use Only
Formula:
C₈H₁₀F₃NO₃ MW: 225.17 g/mol Boiling Pt: 386.6 °C (760 mm Hg) Flash Pt: 187.6 °C Storage Temperature: Ambient |
MDL Number:
MFCD03840837 CAS Number: 126501-70-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...