1-(2-(Trifluoromethyl)pyridin-4-yl)ethanone 97%
Supplier: Ambeed
|
Warning
|
A304385-100MG
A304385-250MG
A304385-1G
A304385-5G
103814-416EA
117
USD
103814-416
103835-582
76827-930
76827-932
76838-274
76838-276
1-(2-(Trifluoromethyl)pyridin-4-yl)ethanone 97%
1-(2-(Trifluoromethyl)pyridin-4-yl)ethanone
Precaución: for Research Use Only
Formula:
C₈H₆F₃NO Storage Temperature: Ambient |
MDL Number:
MFCD11847720 CAS Number: 1060810-86-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...