2'-(Trifluoromethyl)acetophenone 98%

Supplier: Ambeed
Warning

Synonyms: o-(Trifluoromethyl)acetophenone, 1-[2-(Trifluoromethyl)phenyl]ethan-1-one, 1-(2-(Trifluoromethyl)phenyl)ethanone

A159912-100G A159912-1G A159912-25G
103843-528EA 11 USD
103843-528 76997-754 76997-756 77319-432
2'-(Trifluoromethyl)acetophenone 98%
2'-(Trifluoromethyl)acetophenone

Precaución: for Research Use Only

Formula: C₉H₇F₃O
MW: 188.15 g/mol
Boiling Pt: 206.1 °C (760 mm Hg)
Melting Pt: 15…17 °C
Flash Pt: 84 °C
Storage Temperature: Ambient
MDL Number: MFCD00000378
CAS Number: 17408-14-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR