2'-Trifluoromethoxyacetophenone 98%

Supplier: Ambeed
Warning

Synonyms: 1-[2-(Trifluoromethoxy)phenyl]-1-ethanone, 1-(2-(Trifluoromethoxy)phenyl)ethanone

A223017-1G A223017-100G A223017-10G A223017-25G
103819-018EA 5 USD
103819-018 103849-896 77088-514 77261-340 77261-342 77303-640
2'-Trifluoromethoxyacetophenone 98%
2'-Trifluoromethoxyacetophenone

Precaución: for Research Use Only

Formula: C₉H₇F₃O₂
MW: 204.15 g/mol
Boiling Pt: 191.7 °C (760 mm Hg)
Flash Pt: 68.2 °C
Storage Temperature: Ambient
MDL Number: MFCD00236320
CAS Number: 220227-93-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR