2'-(Difluoromethoxy)acetophenone 98%

Supplier: Ambeed
Warning

Synonyms: 1-(2-(Difluoromethoxy)phenyl)ethanone

A266260-1G A266260-250MG
103852-694EA 74 USD
103852-694 77238-640 77238-642
2'-(Difluoromethoxy)acetophenone 98%
2'-(Difluoromethoxy)acetophenone

Precaución: for Research Use Only

Formula: C₉H₈F₂O₂
MW: 186.155 g/mol
Boiling Pt: 225.7 °C (760 mm Hg)
Flash Pt: 87.9 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00236219
CAS Number: 127842-55-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR