2-Acetyl-1H-pyrrole 98%

Supplier: Ambeed
Warning

Synonyms: Methyl-2-pyrrolylketon, Methyl 2-pyrrolyl ketone, 1-(1H-Pyrrol-2-yl)ethanone, 1-(1H-Pyrrol-2-yl)ethan-1-one, 2-Acetylpyrrole

A424265-1G A424265-25G A424265-100G A424265-10G
103827-442EA 5 USD
103827-442 103838-492 76842-134 76842-136 77252-368 77252-370
2-Acetyl-1H-pyrrole 98%
2-Acetyl-1H-pyrrole

Precaución: for Research Use Only

Formula: C₆H₇NO
MW: 109.13 g/mol
Boiling Pt: 220 °C (760 mm Hg)
Melting Pt: 88…93 °C
Flash Pt: 92.4 °C
Storage Temperature: Ambient
MDL Number: MFCD00005220
CAS Number: 1072-83-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR