1-(1H-Indazol-1-yl)ethanone 95%

Supplier: Ambeed
Warning

A506941-1G A506941-5G A506941-250MG
103828-336EA 24 USD
103828-336 103839-438 76843-416 76843-418 77886-302
1-(1H-Indazol-1-yl)ethanone 95%
1-(1H-Indazol-1-yl)ethanone

Precaución: for Research Use Only

Formula: C₉H₈N₂O
Boiling Pt: 306.8 °C (760 mm Hg)
Flash Pt: 139.3 °C
Storage Temperature: Ambient
MDL Number: MFCD16657163
CAS Number: 13436-49-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR