4-Fluoro-α-methylbenzyl bromide 95%

Supplier: Ambeed
Danger

Synonyms: 1-(4-Fluorophenyl)ethyl bromide, 1-Bromo-1-(4-fluorophenyl)ethane, 1-(1-Bromoethyl)-4-fluorobenzene

A175648-10G A175648-1G A175648-25G A175648-5G
103807-072EA 12 USD
103807-072 103845-384 76872-576 77301-394 77301-396 77301-398
4-Fluoro-α-methylbenzyl bromide 95%
4-Fluoro-α-methylbenzyl bromide

Precaución: for Research Use Only

Formula: C₈H₈BrF
MW: 203.052 g/mol
Boiling Pt: 209.6 °C (760 mm Hg)
Flash Pt: 83.3 °C
Storage Temperature: Refrigerator
MDL Number: MFCD11180274
CAS Number: 65130-46-3
UN: 3265
ADR: 8 Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR