1-[(4-Fluorophenyl)carbamoyl]cyclopropanecarboxylic acid 98%

Supplier: Ambeed
Warning

Synonyms: 1-((4-Fluorophenyl)carbamoyl)cyclopropanecarboxylic acid

A192509-100G A192509-10G A192509-250MG A192509-1G A192509-25G A192509-5G
103816-446EA 6 USD
103816-446 103847-054 76974-906 76974-908 76974-910 77084-928 77302-294 77302-296
1-[(4-Fluorophenyl)carbamoyl]cyclopropanecarboxylic acid 98%
1-[(4-Fluorophenyl)carbamoyl]cyclopropanecarboxylic acid

Precaución: for Research Use Only

Formula: C₁₁H₁₀FNO₃
MW: 223.20 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11226313
CAS Number: 849217-48-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR