1-((2-(Diethylamino)ethyl)amino)-4-(hydroxymethyl)-9H-thioxanthen-9-one 95%

Supplier: Ambeed
Warning

A204944-100MG A204944-250MG A204944-25MG A204944-50MG
103792-396EA 41 USD
103792-396 103817-580 77192-446 77192-448 77192-450 77192-452
1-((2-(Diethylamino)ethyl)amino)-4-(hydroxymethyl)-9H-thioxanthen-9-one 95%
1-((2-(Diethylamino)ethyl)amino)-4-(hydroxymethyl)-9H-thioxanthen-9-one

Precaución: for Research Use Only

Formula: C₂₀H₂₄N₂O₂S
MW: 356.48 g/mol
Boiling Pt: 570.5 °C (760 mm Hg)
Flash Pt: 298.9 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00083383
CAS Number: 3105-97-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR