1-(Fmoc-amino)cyclopropanecarboxylic acid 98%

Supplier: Ambeed
Warning

Synonyms: 1-Aminocyclopropane-1-carboxylic acid, N-FMOC protected, Fmoc-ACPC-OH, Fmoc-1-aminocyclopropane-1-carboxylic acid, 1-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)cyclopropanecarboxylic acid

A100290-25G A100290-1G
103795-350EA 27 USD
103795-350 103821-054 76994-434 77296-632
1-(Fmoc-amino)cyclopropanecarboxylic acid 98%
1-(Fmoc-amino)cyclopropanecarboxylic acid

Precaución: for Research Use Only

Formula: C₁₉H₁₇NO₄
MW: 323.348 g/mol
Boiling Pt: 566.9 °C (760 mm Hg)
Melting Pt: 225…226 °C
Flash Pt: 296.7 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00190873
CAS Number: 126705-22-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR