(4,4'-Di-tert-butyl-2,2'-bipyridine)bis[(2-pyridinyl)phenyl]iridium(III) Hexafluorophosphate 97%

Supplier: Ambeed
Warning

Synonyms: [Ir(dtbbpy)(ppy)2]PF6

A334272-1G A334272-100MG A334272-250MG A334272-5G
103825-656EA 148 USD
103825-656 76839-630 77239-812 77239-814 77550-954
(4,4'-Di-tert-butyl-2,2'-bipyridine)bis[(2-pyridinyl)phenyl]iridium(III) Hexafluorophosphate 97%
(4,4'-Di-tert-butyl-2,2'-bipyridine)bis[(2-pyridinyl)phenyl]iridium(III) Hexafluorophosphate

Precaución: for Research Use Only

Formula: C₄₀H₄₀F₆IrN₄P
MW: 913.97 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD27952560
CAS Number: 676525-77-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR