2,2'-Bicinchoninic acid 97%

Supplier: Ambeed
Warning

A216362-1G A216362-25G A216362-5G A216362-250MG A216362-10G
103818-554EA 13 USD
103818-554 103849-274 77087-848 77087-850 77087-852 77249-238 77615-818
2,2'-Bicinchoninic acid 97%
2,2'-Bicinchoninic acid

Precaución: for Research Use Only

Formula: C₂₀H₁₂N₂O₄
Boiling Pt: 626.2 °C (760 mm Hg)
Flash Pt: 332.5 °C
Storage Temperature: Ambient
MDL Number: MFCD00068342
CAS Number: 1245-13-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR