[2,2'-Bipyridine]-6,6'(1H,1'H)-dione 98%

Supplier: Ambeed
Warning

A512644-1G A512644-100MG A512644-10G A512644-250MG
103828-414EA 43 USD
103828-414 103839-514 76843-502 77169-434 77169-436 77169-438
[2,2'-Bipyridine]-6,6'(1H,1'H)-dione 98%
[2,2'-Bipyridine]-6,6'(1H,1'H)-dione

Precaución: for Research Use Only

Formula: C₁₀H₈N₂O₂
MW: 188.18 g/mol
Melting Pt: 325…326 °C
Flash Pt: 275.6 °C
Storage Temperature: Ambient
MDL Number: MFCD18452865
CAS Number: 103505-54-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR