[2,2'-Bipyridine]-4,4'-diyldiphosphonic acid 97%

Supplier: Ambeed
Warning

A306972-100MG A306972-250MG
103814-490EA 360 USD
103814-490 77015-078 77015-080
[2,2'-Bipyridine]-4,4'-diyldiphosphonic acid 97%
[2,2'-Bipyridine]-4,4'-diyldiphosphonic acid

Precaución: for Research Use Only

Formula: C₁₀H₁₀N₂O₆P₂
MW: 316.14 g/mol
Boiling Pt: 741.1 °C (760 mm Hg)
Flash Pt: 402 °C
Storage Temperature: Ambient
MDL Number: MFCD11042490
CAS Number: 194800-56-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR