4,4'-Bis(hydroxymethyl)-2,2'-bipyridine 97%

Supplier: Ambeed
Warning

Synonyms: 4,4'-Bis(hydroxymethyl)-2,2'-bipyridyl, [2,2'-Bipyridine]-4,4'-diyldimethanol, 2,2'-Bipyridine-4,4'-dimethanol

A171053-250MG A171053-25G A171053-1G A171053-5G
103806-582EA 25 USD
103806-582 103844-856 76998-200 76998-202 77301-090 77301-092
4,4'-Bis(hydroxymethyl)-2,2'-bipyridine 97%
4,4'-Bis(hydroxymethyl)-2,2'-bipyridine

Precaución: for Research Use Only

Formula: C₁₂H₁₂N₂O₂
MW: 216.24 g/mol
Boiling Pt: 450.4 °C (760 mm Hg)
Melting Pt: 174 °C
Storage Temperature: Ambient
MDL Number: MFCD06202808
CAS Number: 109073-77-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR