2,2'-Bipyridine-4,4'-dicarboxylic acid 97%

Supplier: Ambeed
Warning

A277149-100G A277149-10G A277149-250MG A277149-1G A277149-25G A277149-5G A277149-500G
103813-336EA 14 USD
103813-336 103853-272 76723-156 76723-158 76723-160 77093-150 77093-152 77093-154 77668-212
2,2'-Bipyridine-4,4'-dicarboxylic acid 97%
2,2'-Bipyridine-4,4'-dicarboxylic acid
Formula: C₁₂H₈N₂O₄
Boiling Pt: 677 °C (760 mm Hg)
Melting Pt: >310 °C
Flash Pt: 363.2 °C
Storage Temperature: Ambient
MDL Number: MFCD00015430
CAS Number: 6813-38-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR