2,2'-Bipyridine-4,4'-dicarbaldehyde 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
[2,2'-Bipyridine]-4,4'-dicarbaldehyde
A166408-250MG
A166408-1G
A166408-5G
A166408-100MG
103806-062EA
137
USD
103806-062
103844-324
77236-134
77320-572
77320-574
77615-382
2,2'-Bipyridine-4,4'-dicarbaldehyde 98%
2,2'-Bipyridine-4,4'-dicarbaldehyde
Precaución: for Research Use Only
Formula:
C₁₂H₈N₂O₂ Boiling Pt: 434.4±45.0 °C (760 mm Hg) Melting Pt: 188 °C Flash Pt: 217.3 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00667770 CAS Number: 99970-84-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...