[2-(2-Hydroxy-ethoxy)-ethyl]-carbamic acid benzyl ester 95%
Supplier: Ambeed
|
Warning
|
A235930-1G
A235930-5G
A235930-100MG
A235930-250MG
A235930-25G
103811-044EA
10
USD
103811-044
103850-814
77089-836
77089-838
77152-998
77153-000
77568-282
[2-(2-Hydroxy-ethoxy)-ethyl]-carbamic acid benzyl ester 95%
[2-(2-Hydroxy-ethoxy)-ethyl]-carbamic acid benzyl ester
Precaución: For research use only.
Formula:
C₁₂H₁₇NO₄ MW: 239.26 g/mol Boiling Pt: 420.6 °C (760 mm Hg) Flash Pt: 208.2 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD15143230 CAS Number: 145881-74-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...