[1,2,4]Triazolo[1,5-a]pyridine-6-boronic Acid Pinacol Ester 97%

Supplier: Ambeed
Warning

A294777-5G A294777-100MG A294777-250MG
103814-026EA 159 USD
103814-026 103835-166 76837-806 76875-558 76875-560
[1,2,4]Triazolo[1,5-a]pyridine-6-boronic Acid Pinacol Ester 97%
[1,2,4]Triazolo[1,5-a]pyridine-6-boronic Acid Pinacol Ester

Precaución: for Research Use Only

Formula: C₁₂H₁₆BN₃O₂
MW: 245.09 g/mol
Melting Pt: 109…114 °C
Storage Temperature: Refrigerator
MDL Number: MFCD16659797
CAS Number: 1160790-18-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR