[1,2,4]Triazolo[1,5-a]pyridin-7-ol 95%, Ambeed.Inc

Supplier: Ambeed
Warning

A325033-1G A325033-5G A325033-250MG A325033-100MG A325033-10G A325033-25G
103825-388EA 32 USD
103825-388 103836-332 76839-256 76839-258 77167-342 77683-967 77892-668 77892-669
[1,2,4]Triazolo[1,5-a]pyridin-7-ol 95%, Ambeed.Inc
[1,2,4]Triazolo[1,5-a]pyridin-7-ol

Precaución: For research use only.

Formula: C₆H₅N₃O
Storage Temperature: Refrigerator
MDL Number: MFCD22394734
CAS Number: 1033810-70-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR