[1,1'-Bis(diphenylphosphino)ferrocene]nickel(II) chloride 98%

Supplier: Ambeed
Danger

Synonyms: Dichloro[1,1'-bis(diphenylphosphino)ferrocene]nickel(II), [1,1'-Bis(diphenylphosphino)ferrocene]dichloronickel(II), [1,1’-Bis(diphenylphosphino)ferrocene]dichloronickel(II)

A606847-100G A606847-10G A606847-25G A606847-1G A606847-5G
103840-144EA 6 USD
103840-144 76819-444 76819-446 76844-310 77307-930 77307-932
[1,1'-Bis(diphenylphosphino)ferrocene]nickel(II) chloride 98%
[1,1'-Bis(diphenylphosphino)ferrocene]nickel (II) chloride

Precaución: for Research Use Only

Formula: C₃₄H₂₈Cl₂FeNiP₂
MW: 683.98 g/mol
Melting Pt: 285 °C
Storage Temperature: Ambient
MDL Number: MFCD00270284
CAS Number: 67292-34-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR