4-Biphenylboronic acid 98%

Supplier: Ambeed
Warning

Synonyms: [1,1'-Biphenyl]-4-ylboronic acid

A105961-100G A105961-10G A105961-25G A105961-500G A105961-5G
103796-976EA 5 USD
103796-976 103822-724 76796-518 76796-520 76796-522 76796-524 77283-414 77283-416
4-Biphenylboronic acid 98%
4-Biphenylboronic acid

Precaución: for Research Use Only

Formula: C₁₂H₁₁BO₂
Boiling Pt: 385.5 °C (760 mm Hg)
Melting Pt: 232…245 °C
Flash Pt: 186.9 °C
Storage Temperature: Ambient
MDL Number: MFCD00093311
CAS Number: 5122-94-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR