Biphenyl-4-ol 97%

Supplier: Ambeed
Danger

Synonyms: [1,1'-Biphenyl]-4-ol, 4-Hydroxydiphenyl, 4-Phenylphenol, p-Hydroxybiphenyl, 4-Hydroxybiphenyl

A107909-100G A107909-25G A107909-500G A107909-1000G
103807-978EA 93 USD
103807-978 76869-528 76869-530 76869-532 77283-662
Biphenyl-4-ol 97%
Biphenyl-4-ol

Precaución: for Research Use Only

Formula: C₁₂H₁₀O
MW: 170.21 g/mol
Boiling Pt: 321 °C
Melting Pt: 164…166 °C
Flash Pt: 165.6 °C
Storage Temperature: Ambient
MDL Number: MFCD00002347
CAS Number: 92-69-3
UN: 3077
ADR: 9,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR