4-Phenylbenzoic acid 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
Biphenyl-4-carboxylic acid, [1,1'-Biphenyl]-4-carboxylic acid, 4-Biphenylcarboxylic acid
A121028-1000G
A121028-100G
A121028-10G
A121028-25G
A121028-500G
A121028-5G
76718-526EA
164.44
USD
76718-526
76718-528
76718-530
76718-532
76718-534
77297-808
4-Phenylbenzoic acid 98%
4-Phenylbenzoic acid
Precaución: for Research Use Only
Formula:
C₁₃H₁₀O₂ MW: 198.22 g/mol Boiling Pt: 372.6 °C (760 mm Hg) Melting Pt: 220…225 °C Storage Temperature: Ambient |
MDL Number:
MFCD00002553 CAS Number: 92-92-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...