[1,1'-Biphenyl]-4-carboximidamide hydrochloride 97%

Supplier: Ambeed
Warning

A100472-100MG A100472-250MG A100472-1G
103795-458EA 12 USD
103795-458 103821-152 77008-272 77008-274 77296-700
[1,1'-Biphenyl]-4-carboximidamide hydrochloride 97%
[1,1'-Biphenyl]-4-carboximidamide hydrochloride

Precaución: for Research Use Only

Formula: C₁₃H₁₃ClN₂
MW: 232.71 g/mol
Melting Pt: 248…251 °C
Storage Temperature: Ambient
MDL Number: MFCD04114455
CAS Number: 111082-23-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR