4-Phenylbenzonitrile 98%

Supplier: Ambeed
Danger

Synonyms: 4-Biphenylcarbonitrile, 4-Cyanobiphenyl, p-Phenylbenzonitrile, [1,1'-Biphenyl]-4-carbonitrile

A530286-10G A530286-1G A530286-25G A530286-5G
103839-672EA 6 USD
103839-672 76726-290 76726-292 76843-690 76843-692
4-Phenylbenzonitrile 98%
4-Phenylbenzonitrile

Precaución: for Research Use Only

Formula: C₁₃H₉N
MW: 179.22 g/mol
Boiling Pt: 332.3 °C (760 mm Hg)
Melting Pt: 85…87 °C
Flash Pt: 110 °C
Storage Temperature: Ambient
MDL Number: MFCD00001821
CAS Number: 2920-38-9
UN: 3439
ADR: 6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR