4-Phenylbenzaldehyde 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
Biphenyl-4-carbaldehyde, [1,1'-Biphenyl]-4-carbaldehyde, 4-Biphenylcarboxaldehyde, Biphenyl-4-carboxaldehyde, p-Biphenylylaldehyde, 4-Biphenylylaldehyde, 4-Biphenylcarbaldehyde
A197007-100G
A197007-10G
A197007-500G
A197007-25G
A197007-5G
103847-500EA
7
USD
103847-500
77274-664
77274-666
77274-668
77302-544
77302-546
4-Phenylbenzaldehyde 98%
4-Phenylbenzaldehyde
Precaución: for Research Use Only
Formula:
C₁₃H₁₀O MW: 182.22 g/mol Boiling Pt: 326.6 °C (760 mm Hg) Melting Pt: 57…61 °C Flash Pt: 186.1 °C Storage Temperature: Ambient |
MDL Number:
MFCD00006947 CAS Number: 3218-36-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...