4,4'-Biphenyldiboronic acid 97%

Supplier: Ambeed
Warning

Synonyms: [1,1'-Biphenyl]-4,4'-diyldiboronic acid

A151518-250MG A151518-5G A151518-1G
103804-266EA 7 USD
103804-266 103842-418 77095-824 77299-884 77317-990
4,4'-Biphenyldiboronic acid 97%
4,4'-Biphenyldiboronic acid

Precaución: for Research Use Only

Formula: C₁₂H₁₂B₂O₄
MW: 241.85 g/mol
Boiling Pt: 505.9 °C (760 mm Hg)
Melting Pt: 300 °C
Flash Pt: 259.7 °C
Storage Temperature: Ambient
MDL Number: MFCD00151795
CAS Number: 4151-80-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR