Biphenyl-4,4'-dicarboxylic acid 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
4,4'-Biphenyldicarboxylic acid, [1,1'-Biphenyl]-4,4'-dicarboxylic acid
A157152-100G
A157152-10G
A157152-25G
A157152-5G
103843-174EA
5
USD
103843-174
77273-166
77273-168
77318-994
77318-996
Biphenyl-4,4'-dicarboxylic acid 97%
Biphenyl-4,4'-dicarboxylic acid
Precaución: for Research Use Only
Formula:
C₁₄H₁₀O₄ MW: 242.23 g/mol Melting Pt: >300 °C Flash Pt: 252.9 °C Storage Temperature: Ambient |
MDL Number:
MFCD00002554 CAS Number: 787-70-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...