4,4'-Biphenyldicarboxaldehyde 95%

Supplier: Ambeed
Warning

Synonyms: [1,1'-Biphenyl]-4,4'-dicarbaldehyde, 4,4'-Diformylbiphenyl

A161693-25G A161693-5G A161693-100G
103843-760EA 14 USD
103843-760 77319-752 77319-754 77671-292
4,4'-Biphenyldicarboxaldehyde 95%
4,4'-Biphenyldicarboxaldehyde

Precaución: for Research Use Only

Formula: C₁₄H₁₀O₂
MW: 210.23 g/mol
Boiling Pt: 391.7 °C (760 mm Hg)
Melting Pt: 144…147 °C
Flash Pt: 146.8 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00016714
CAS Number: 66-98-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR