3-Biphenylboronic acid 98%

Supplier: Ambeed
Warning

Synonyms: [1,1'-Biphenyl]-3-ylboronic acid

A118718-100G A118718-10G A118718-25G A118718-1G A118718-5G
103799-100EA 10 USD
103799-100 76797-244 76797-246 76797-248 77311-772 77311-774
3-Biphenylboronic acid 98%
3-Biphenylboronic acid

Precaución: for Research Use Only

Formula: C₁₂H₁₁BO₂
MW: 198.03 g/mol
Boiling Pt: 411 °C (760 mm Hg)
Melting Pt: 193…198 °C
Flash Pt: 202.3 °C
Storage Temperature: Ambient
MDL Number: MFCD01318102
CAS Number: 5122-95-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR