3-Phenylbenzoic acid 98%

Supplier: Ambeed
Warning

Synonyms: Biphenyl-3-carboxylic acid, 3-Biphenylcarboxylic acid, 1,1'-Biphenyl-3-carboxylic acid, [1,1'-Biphenyl]-3-carboxylic acid

A109729-10G A109729-250MG A109729-25G A109729-1G A109729-5G
103797-554EA 9 USD
103797-554 103823-360 76718-032 76718-034 76718-036 77310-168 77310-170
3-Phenylbenzoic acid 98%
3-Phenylbenzoic acid

Precaución: for Research Use Only

Formula: C₁₃H₁₀O₂
MW: 198.22 g/mol
Melting Pt: 164…168 °C
Flash Pt: 177.3 °C
Storage Temperature: Ambient
MDL Number: MFCD00045846
CAS Number: 716-76-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR