[1,1'-Biphenyl]-3,3',4,4'-tetracarboxylic acid 97%

Supplier: Ambeed
Warning

A120282-250MG A120282-25G A120282-1G A120282-5G
103799-908EA 17 USD
103799-908 77026-352 77026-354 77297-746 77297-748
[1,1'-Biphenyl]-3,3',4,4'-tetracarboxylic acid 97%
[1,1'-Biphenyl]-3,3',4,4'-tetracarboxylic acid

Precaución: for Research Use Only

Formula: C₁₆H₁₀O₈
MW: 330.25 g/mol
Boiling Pt: 673.3 °C (760 mm Hg)
Melting Pt: 280 °C
Flash Pt: 375 °C
Storage Temperature: Ambient
MDL Number: MFCD00496632
CAS Number: 22803-05-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR