2-Phenylbenzoic acid 98%

Supplier: Ambeed
Warning

Synonyms: Biphenyl-2-carboxylic acid, 2-Biphenylcarboxylic acid, [1,1'-Biphenyl]-2-carboxylic acid

A348320-100G A348320-10G A348320-500G A348320-25G A348320-5G
103836-980EA 5 USD
103836-980 76724-414 76724-416 76724-418 76840-058 77306-596
2-Phenylbenzoic acid 98%
2-Phenylbenzoic acid

Precaución: for Research Use Only

Formula: C₁₃H₁₀O₂
MW: 198.22 g/mol
Melting Pt: 111…113 °C
Flash Pt: 343 °C
Storage Temperature: Ambient
MDL Number: MFCD00002463
CAS Number: 947-84-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR