5'-m-Terphenylboronic acid 98%

Supplier: Ambeed
Warning

Synonyms: [1,1':3',1''-Terphenyl]-5'-ylboronic acid

A288541-100G A288541-10G A288541-25G A288541-5G
103813-800EA 6 USD
103813-800 103834-942 76827-666 76827-668 76837-504 76837-506
5'-m-Terphenylboronic acid 98%
5'-m-Terphenylboronic acid

Precaución: for Research Use Only

Formula: C₁₈H₁₅BO₂
MW: 274.13 g/mol
Boiling Pt: 513 °C (760 mm Hg)
Melting Pt: 292…296 °C
Flash Pt: 264.1 °C
Storage Temperature: Ambient
MDL Number: MFCD09953491
CAS Number: 128388-54-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR