(R)-(-)-1-[(S)-2-(Dicyclohexylphosphino)ferrocenyl]ethyldi-t-butylphosphine 97%

Supplier: Ambeed
Warning

Synonyms: (2R)-1-[(1R)-1-[Bis(1,1-dimethylethyl)phosphino]ethyl]-2-(dicyclohexylphosphino)ferrocene, [(R)-1-[(R)-2-(Dicyclohexylphosphino)ferrocenyl]ethyl]di-tert-butylphosphine

A235385-100MG A235385-10G A235385-250MG
103811-008EA 23 USD
103811-008 103850-776 76874-320 76874-322 76874-324
(R)-(-)-1-[(S)-2-(Dicyclohexylphosphino)ferrocenyl]ethyldi-t-butylphosphine 97%
(R)-(-)-1-[(S)-2-(Dicyclohexylphosphino)ferrocenyl]ethyldi-t-butylphosphine

Precaución: for Research Use Only

Formula: C₃₂H₅₂FeP₂
Storage Temperature: Refrigerator
MDL Number: MFCD08543435
CAS Number: 158923-11-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR