(Z)-tert-Butyl-2-(((1-(2-aminothiazol-4-yl)-2-(benzo[d]thiazol-2-ylthio)-2-oxoethylidene)amino)oxy)-2-methylpropanoate 97%

Supplier: Ambeed
Danger

Synonyms: (Z)-tert-Butyl 2-(((1-(2-aminothiazol-4-yl)-2-(benzo[d]thiazol-2-ylthio)-2-oxoethylidene)amino)oxy)-2-methylpropanoate

A217206-10G A217206-1G A217206-5G
103849-336EA 8 USD
103849-336 76975-734 76975-736 77087-954
(Z)-tert-Butyl-2-(((1-(2-aminothiazol-4-yl)-2-(benzo[d]thiazol-2-ylthio)-2-oxoethylidene)amino)oxy)-2-methylpropanoate 97%
(Z)-tert-Butyl-2-(((1-(2-aminothiazol-4-yl)-2-(benzo[d]thiazol-2-ylthio)-2-oxoethylidene)amino)oxy)-2-methylpropanoate

Precaución: for Research Use Only

Formula: C₂₀H₂₂N₄O₄S₃
MW: 478.61 g/mol
Boiling Pt: 621.6 °C (760 mm Hg)
Melting Pt: 138…140 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00071548
CAS Number: 89604-92-2
UN: 1325
ADR: 4.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR