(Z)-5-((5-Fluoro-2-oxoindolin-3-ylidene)methyl)-2,4-dimethyl-1H-pyrrole-3-carboxylic acid 97%

Supplier: Ambeed
Warning

A114706-250MG A114706-25G A114706-1G A114706-5G
103798-456EA 74 USD
103798-456 103824-300 77026-014 77026-016 77311-114 77311-116
(Z)-5-((5-Fluoro-2-oxoindolin-3-ylidene)methyl)-2,4-dimethyl-1H-pyrrole-3-carboxylic acid 97%
(Z)-5-((5-Fluoro-2-oxoindolin-3-ylidene)methyl)-2,4-dimethyl-1H-pyrrole-3-carboxylic acid

Precaución: for Research Use Only

Formula: C₁₆H₁₃FN₂O₃
MW: 300.28 g/mol
Boiling Pt: 588.4 °C (760 mm Hg)
Flash Pt: 309.683 °C
Storage Temperature: Ambient
MDL Number: MFCD11521323
CAS Number: 356068-93-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR